Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl isobutyrate, 99%

Catalog No. p-164363
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 547-63-7 | C5H10O2 | 102.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 547-63-7
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00008914
InChI Key BHIWKHZACMWKOJ-UHFFFAOYSA-N
Synonym methyl isobutyrate, methyl 2-methylpropionate, methyl isobutanoate, propanoic acid, 2-methyl-, methyl ester, isobutyric acid, methyl ester, methylisobutyrate, isobutyric acid methyl ester, poly d-lactide, methyl isobutyrate natural, methylester kyseliny isomaselne
PubChem CID 11039
ChEBI CHEBI:73689
IUPAC Name methyl 2-methylpropanoate
SMILES CC(C)C(=O)OC

Specifications

Acidity 1mg KOH/g max.
Melting Point -85°C to 84°C
Color Colorless
Density 0.8900g/mL
Boiling Point 90°C
Flash Point 1°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3790 to 1.384
Linear Formula (CH3)2CHCO2CH3
Quantity 100 g
Beilstein 02, 290
Specific Gravity 0.89
Merck Index 15, 6160
Solubility Information Solubility in water: slightly soluble. Other solubilities: miscible with alcohol and ether
Viscosity < 30 mPa.s
Formula Weight 102.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl isobutyrate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 208-929-5
RTECSNumber NQ5425000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.