Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl isonicotinate, 98%

Catalog No. AAB2326322
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 2459-09-8 | C7H7NO2 | 137.14 g/mol

Methyl Isonicotinate induces increased walking and take-off behavior in western flower thrips, Frankliniella occidentalis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl Isonicotinate induces increased walking and take-off behavior in western flower thrips, Frankliniella occidentalis.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2459-09-8
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00006427
InChI Key OLXYLDUSSBULGU-UHFFFAOYSA-N
Synonym methyl isonicotinate, isonicotinic acid methyl ester, 4-pyridinecarboxylic acid, methyl ester, 4-picolinic acid methyl ester, 4-methoxycarbonylpyridine, methylisonicotinate, methyl 4-pyridinecarboxylate, 4-carbomethoxypyridine, isonicotinic acid, methyl ester, i-nicotinic acid, methyl ester
PubChem CID 227085
IUPAC Name methyl pyridine-4-carboxylate
SMILES COC(=O)C1=CC=NC=C1

Specifications

Melting Point 8°C to 8.5°C
Density 1.016
Boiling Point 207°C to 209°C
Flash Point 82°C (180°F)
Odor Characteristic
Refractive Index 1.5135
Quantity 100 g
Solubility Information Slightly soluble in water.
Formula Weight 137.14
Percent Purity 98%
Chemical Name or Material Methyl isonicotinate
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H319-H227
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 219-456-8
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.