Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl isothiocyanate, 98%

Catalog No. aaa1175714
Click to view available options
Quantity:
25 g

CAS: 556-61-6 | C2H3NS | 73.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 556-61-6
Molecular Formula C2H3NS
Molecular Weight (g/mol) 73.11
MDL Number MFCD00004818,MFCD03457480
InChI Key LGDSHSYDSCRFAB-UHFFFAOYSA-N
Synonym methyl isothiocyanate, isothiocyanatomethane, methane, isothiocyanato, trapex, methyl mustard oil, methyl mustard, mitc, mencs, methylisothiocyanate, trapexide
PubChem CID 11167
ChEBI CHEBI:78337
IUPAC Name isothiocyanatomethane
SMILES CN=C=S

Specifications

Melting Point 31°C to 36°C
Density 1.069
Boiling Point 117°C to 118°C
Flash Point 30°C (86°F)
Linear Formula CH3NCS
Quantity 25 g
UN Number UN2477
Beilstein 605319
Sensitivity Moisture sensitive
Merck Index 14,6092
Formula Weight 73.12
Percent Purity 98%
Physical Form Fused Solid
Chemical Name or Material Methyl isothiocyanate
Show More Show Less
Hazard Category H301-H310-H314-H317-H335
Hazard Statement GHS H Statement
H301-H331-H314-H317
Toxic if swallowed.
Toxic if inhaled.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Precautionary Statement P260-P262-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: METHYL ISOTHIOCYANATE
EINECSNumber 209-132-5
RTECSNumber PA9625000
TSCA Yes
storageNote1 2°C to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.