Alfa Aesar™ Methyl isoxazole-5-carboxylate, 97%

Manufacturer: Alfa Aesar™ B2516903

Quantity 1g

 View more versions of this product

Catalog No. AAB2516903

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 15055-81-9
    Molecular Formula C5H5NO3
    Formula Weight 127.1
    MDL Number MFCD01570794
    Synonym Isoxazole-5-carboxylic acid methyl ester
    Chemical Name or Material Methyl isoxazole-5-carboxylate

    Specifications

    Formula Weight 127.1
    Molecular Formula C5H5NO3
    MDL Number MFCD01570794
    CAS 15055-81-9
    Synonym Isoxazole-5-carboxylic acid methyl ester
    Melting Point 48° to 50°C
    Quantity 1g
    Chemical Name or Material Methyl isoxazole-5-carboxylate
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit