Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl isoxazole-5-carboxylate, 97%

Catalog No. AAB2516906
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g

CAS: 15055-81-9 | C5H5NO3 | 127.099 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15055-81-9
Molecular Formula C5H5NO3
Molecular Weight (g/mol) 127.099
MDL Number MFCD01570794
InChI Key ILPCPKZAAQXHKL-UHFFFAOYSA-N
Synonym methyl isoxazole-5-carboxylate, 5-isoxazolecarboxylic acid, methyl ester, isoxazole-5-carboxylic acid methyl ester, acmc-209wih, methyl 5-isoxazolcarboxylate, methyl 5-isoxazolcarboxylate;, methylisoxazole-5-carboxylate, 5-methoxycarbonyl-1,2-oxazol, 5-isoxazol-carbonsaeure-methylester;, isoxazol-5-carbonsaeure-methylester;
PubChem CID 2736892
IUPAC Name methyl 1,2-oxazole-5-carboxylate
SMILES COC(=O)C1=CC=NO1

Specifications

Melting Point 48°C to 50°C
Quantity 5 g
Formula Weight 127.1
Percent Purity 97%
Chemical Name or Material Methyl isoxazole-5-carboxylate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.