Alfa Aesar™ Methyl methoxyacetate, 99%

Manufacturer: Alfa Aesar™ B2252836

Quantity 500g

 View more versions of this product

Catalog No. AAB2252836

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6290-49-9
    Molecular Formula CH3OCH2CO2CH3
    Formula Weight 104.11
    MDL Number MFCD00008451
    UN Number UN3272
    Beilstein 1742944
    Synonym Methoxyacetic acid methyl ester
    Chemical Name or Material Methyl methoxyacetate

    Specifications

    Formula Weight 104.11
    TSCA TSCA
    Molecular Formula CH3OCH2CO2CH3
    MDL Number MFCD00008451
    UN Number UN3272
    Beilstein 1742944
    Synonym Methoxyacetic acid methyl ester
    Boiling Point 129° to 130°C
    Quantity 500g
    Flash Point 35°C (95°F)
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226
    Flammable liquid and vapour.
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
    RTECS Number AI8910000
    CAS 6290-49-9
    Chemical Name or Material Methyl methoxyacetate
    Assay 99%

    Description

    Applications
    Methyl methoxyacetate is used in the preparation of 4-hydroxy-2-mercapto-5-methoxypyrimidine (II).

    Solubility
    Soluble in water.

    Notes
    Keep container tightly closed. Store away from oxidizing agents.

    GHS H Statement
    H226
    Flammable liquid and vapour.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit