Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl myristate, 99%

Catalog No. p-165469
Click to view available options
Quantity:
2.5 mL
100 mL
500 mL
Packaging:
Glass bottle

CAS: 124-10-7 | C15H30O2 | 242.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-10-7
Molecular Formula C15H30O2
Molecular Weight (g/mol) 242.40
MDL Number MFCD00008983
InChI Key ZAZKJZBWRNNLDS-UHFFFAOYSA-N
Synonym methyl myristate, tetradecanoic acid, methyl ester, uniphat a50, metholeneat 2495, myristic acid methyl ester, methyl n-tetradecanoate, myristic acid, methyl ester, methyl myristylate, tetradecanoic acid methyl ester, myristic acid, methyl ester 8ci
PubChem CID 31284
IUPAC Name methyl tetradecanoate
SMILES CCCCCCCCCCCCCC(=O)OC

Specifications

Acidity 1mg KOH/g max.
Melting Point 18.4°C
Color Colorless to Yellow
Density 0.8630g/mL
Boiling Point 323.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4340 to 1.4380
Linear Formula CH3(CH2)12CO2CH3
Quantity 2.5 mL
Beilstein 02, II, 326
Specific Gravity 0.863
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in ethanol
Formula Weight 242.4
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl myristate, 99%
Show More Show Less
EINECSNumber 204-680-1
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.