Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl nicotinate, 99%

Catalog No. p-7027278
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2390822 100 g
AAB2390830 250 g
AAB2390836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2390822 Supplier Thermo Scientific Chemicals Supplier No. B2390822
Only null left
Add to Cart

CAS: 93-60-7 | C7H7NO2 | 137.138 g/mol

Used as a rubefacient, topical vasodilator used to test the efficacy of skin protection compounds (i.e. products designed to provide barriers to chemical warfare agents) and used as a flavoring agent. It is a useful synthetic intermediate. It can be used to prepare biarylcarboxamide inhibitors of phosphodiesterase IV and tumor necrosis factor-α in treatment of rheumatoid arthritis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a rubefacient, topical vasodilator used to test the efficacy of skin protection compounds (i.e. products designed to provide barriers to chemical warfare agents) and used as a flavoring agent. It is a useful synthetic intermediate. It can be used to prepare biarylcarboxamide inhibitors of phosphodiesterase IV and tumor necrosis factor-α in treatment of rheumatoid arthritis.

Solubility
Soluble in water at 20°C 48 g/L. Soluble in alcohol, chloroform and ethyl acetate. Solubility in methanol (almost transparency).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-60-7
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.138
MDL Number MFCD00006388
InChI Key YNBADRVTZLEFNH-UHFFFAOYSA-N
Synonym methyl nicotinate, nikomet, methylnicotinate, nicotinic acid methyl ester, nicometh, methyl 3-pyridinecarboxylate, methyl-nicotinate, 3-pyridinecarboxylic acid, methyl ester, heat spray, nicotinic acid, methyl ester
PubChem CID 7151
IUPAC Name methyl pyridine-3-carboxylate
SMILES COC(=O)C1=CN=CC=C1

Specifications

Melting Point 40°C to 44°C
Boiling Point 208°C to 210°C
Flash Point 95°C (203°F)
Odor Slightly Sweetish
Quantity 100 g
Beilstein 113951
Merck Index 14,6100
Solubility Information Soluble in water at 20°C 48g/L. Soluble in alcohol,chloroform and ethyl acetate. Solubility in methanol (almost transparency).
Formula Weight 137.14
Percent Purity 99%
Chemical Name or Material Methyl nicotinate
Show More Show Less
Hazard Category H315-H319-H500
Hazard Statement GHS H Statement
H319-H317
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 202-261-8
RTECSNumber QT1925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.