Learn More
Methyl nicotinate, 99%
Description
Used as a rubefacient, topical vasodilator used to test the efficacy of skin protection compounds (i.e. products designed to provide barriers to chemical warfare agents) and used as a flavoring agent. It is a useful synthetic intermediate. It can be used to prepare biarylcarboxamide inhibitors of phosphodiesterase IV and tumor necrosis factor-α in treatment of rheumatoid arthritis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Used as a rubefacient, topical vasodilator used to test the efficacy of skin protection compounds (i.e. products designed to provide barriers to chemical warfare agents) and used as a flavoring agent. It is a useful synthetic intermediate. It can be used to prepare biarylcarboxamide inhibitors of phosphodiesterase IV and tumor necrosis factor-α in treatment of rheumatoid arthritis.
Solubility
Soluble in water at 20°C 48 g/L. Soluble in alcohol, chloroform and ethyl acetate. Solubility in methanol (almost transparency).
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Melting Point | 40°C to 44°C |
| Boiling Point | 208°C to 210°C |
| Flash Point | 95°C (203°F) |
| Odor | Slightly Sweetish |
| Quantity | 100 g |
| Beilstein | 113951 |
| Merck Index | 14,6100 |
| Solubility Information | Soluble in water at 20°C 48g/L. Soluble in alcohol,chloroform and ethyl acetate. Solubility in methanol (almost transparency). |
| Formula Weight | 137.14 |
| Percent Purity | 99% |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H500 |
| Hazard Statement | GHS H Statement H319-H317 Causes serious eye irritation. May cause an allergic skin reaction. |
| Precautionary Statement | P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362 |
| EINECSNumber | 202-261-8 |
| RTECSNumber | QT1925000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.