Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl nonanoate, ≥99.8% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7188014
Click to view available options
Quantity:
1 mL
5 mL
Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 1731-84-6
Density 0.875 g/mL (at 25°C)
Flash Point 87°C - Closed Cup
Shipping Condition Ambient
Physical Form Neat
Linear Formula CH3(CH2)7 COOCH3
MDL Number MFCD00009569
Boiling Point 213°C to 214°C
Synonym Methyl pelargonate; Nonanoic acid methyl ester
Type Analytical Standard
Quantity 1 mL
Formula Weight 172.26
Percent Purity ≥99.8% (GC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.