Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl oleate, 96%

Catalog No. p-7030661
Click to view available options
Quantity:
25 g
100 g

CAS: 112-62-9 | C19H36O2 | 296.495 g/mol

Methyl oleate is used as a chromatographic reference standard in biochemical research. It is also used as an intermediate for detergents, emulsifiers, wetting agents, stabilizers, textile treatments, plasticizers for duplicating inks, rubbers and waxes. It finds application as lubricants and lubricant additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl oleate is used as a chromatographic reference standard in biochemical research. It is also used as an intermediate for detergents, emulsifiers, wetting agents, stabilizers, textile treatments, plasticizers for duplicating inks, rubbers and waxes. It finds application as lubricants and lubricant additives.

Solubility
Miscible with alcohol, ether and chloroform. Immiscible with water.

Notes
Air and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-62-9
Molecular Formula C19H36O2
Molecular Weight (g/mol) 296.495
MDL Number MFCD00009578
InChI Key QYDYPVFESGNLHU-KHPPLWFESA-N
Synonym methyl oleate, oleic acid methyl ester, methyl cis-9-octadecenoate, oleic acid, methyl ester, edenor metio5, methyl z-9-octadecenoate, methyl-cis-oleate, exceparl m-ol, emery, oleic acid ester, esterol 112
PubChem CID 5364509
ChEBI CHEBI:27542
IUPAC Name methyl (Z)-octadec-9-enoate
SMILES CCCCCCCCC=CCCCCCCCC(=O)OC

Specifications

Density 0.876
Boiling Point 168°C to 170°C (2 mmHg)
Flash Point 163°C (325°F)
Refractive Index 1.452
Linear Formula CH3(CH2)7CH=CH(CH2)7CO2CH3
Quantity 25 g
Beilstein 1727037
Merck Index 14,6828
Solubility Information Miscible with alcohol,ether and chloroform. Immiscible with water.
Formula Weight 296.5
Percent Purity 96%
Chemical Name or Material Methyl oleate
Show More Show Less
EINECSNumber 203-992-5
RTECSNumber RK0895000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.