Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Orange Indicator, 0.01% w/v, Reagents

Catalog No. p-200021858
Click to view available options
Quantity:
100 mL
500 mL

Methyl Orange Indicator, 0.01% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Methyl Orange Indicator, 0.01% w/v
CAS 7732-18-5, 547-58-0
Boiling Point 100°C
Molecular Formula C14H14N3NaO3S/H2O
Synonym Orange III, Helianthine
Percent Purity 0.01%
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.001
Color Orange
Quantity 100 mL
Physical Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.