Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methyl Orange Indicator, 0.05% w/v, Reagents
Click to view available options
Quantity:
120 mL
500 mL
1 L
4 L
Specifications
Specifications
| Chemical Name or Material | Methyl Orange Indicator, 0.05% w/v |
| CAS | 7732-18-5, 547-58-0 |
| Boiling Point | 100°C |
| Molecular Formula | C14H14N3NaO3S/H2O |
| Synonym | Orange III, Helianthine |
| Percent Purity | 0.05% |
| Grade | Reagent |
| Packaging | Poly Bottle |
| Specific Gravity | 1.001 |
| Color | Orange |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction