Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Orange, Reagent, ACS, Spectrum™ Chemical
SDP

Catalog No. 18604435
Click to view available options
Quantity:
25 g
125 g
500 g
5 kg
6 x 125 g
6 x 500 g
25 kg
Packaging:
Amber Glass Bottle

ME160, 547-58-0, C14H14N3O3SNa

Spectrum™ Chemical Methyl Orange, Reagent, ACS is most typically used as an indicator in titrations due to the fact of its color change. The color changes happen in acidic conditions and usually moves from red to orange and eventually to yellow. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 547-58-0
Molecular Formula C14H14N3O3S
Molecular Weight (g/mol) 304.34
InChI Key IETWCRRCPURZOC-UHFFFAOYSA-M
IUPAC Name 4-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzene-1-sulfonate
SMILES CN(C)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)S([O-])(=O)=O

Specifications

CAS Max % 100%
UN Number UN3143
Grade ACS
Packaging Amber Glass Bottle
Color Orange to Yellow
Quantity 125 g
Chemical Name or Material Methyl Orange
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.