Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl p-toluate, 99%

Catalog No. AC414965000
Click to view available options
Quantity:
100 g
250 g
500 g
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic drum

CAS: 99-75-2 | C9H10O2 | 150.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-75-2
MDL Number MFCD00008441
InChI Key QSSJZLPUHJDYKF-UHFFFAOYSA-N
Synonym methyl p-toluate, methyl-p-toluate, p-carbomethoxytoluene, methyl 4-toluate, 4-methylbenzoic acid methyl ester, methyl p-methylbenzoate, benzoic acid, 4-methyl-, methyl ester, 4-methoxycarbonyl toluene, p-toluic acid, methyl ester, methyl p-toluenecarboxylate
PubChem CID 7455
IUPAC Name methyl 4-methylbenzoate
SMILES CC1=CC=C(C=C1)C(=O)OC

Specifications

Density 1.0600g/mL
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C9H10O2
Linear Formula CH3C6H4COOCH3
Quantity 500 g
Molecular Weight (g/mol) 150.177
Formula Weight 150.18
Chemical Name or Material Methyl p-toluate, 99%
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-784-1
RTECSNumber XU2150000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.