Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methyl palmitate, ≥99.0% (GC), MilliporeSigma™ Supelco™
Click to view available options
Quantity:
1 g
5 g
Description
Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications
Specifications
CAS | 112-39-0 |
Density | 0.852 g/mL (at 25°C) |
Flash Point | 113.0°C - Closed Cup |
Shipping Condition | Ambient |
Physical Form | Neat |
Linear Formula | CH3(CH2)14 CO2 CH3 |
MDL Number | MFCD00008994 |
Melting Point | 32°C to 35°C |
Boiling Point | 185°C (10 mmHg) |
Synonym | n-Hexadecanoic acid methyl ester; Methyl hexadecanoate; Palmitic acid methyl ester |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction