Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl palmitate, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7188027 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
1 g
5 g

Methyl palmitate is a fatty acid methyl ester, which shows potential anti-inflammatory and antifibrotic effects by preventing bleomycin-induced lung inflammation and inhibiting the nuclear factor kappa-β pathway(NF-κB).

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 112-39-0
Density 0.852 g/mL (at 25°C)
Flash Point 113.0°C - Closed Cup
Shipping Condition Ambient
Physical Form Neat
Linear Formula CH3(CH2)14 CO2 CH3
MDL Number MFCD00008994
Melting Point 32°C to 35°C
Boiling Point 185°C (10 mmHg)
Synonym n-Hexadecanoic acid methyl ester; Methyl hexadecanoate; Palmitic acid methyl ester
Type Analytical Standard
Quantity 1 g
Formula Weight 270.45
Percent Purity ≥99.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.