Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl pentadecanoate, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111015326 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
1 mL
5 mL
25 mL

Catalog No. 11-101-5326

Supplier: MilliporeSigma Supelco 7656025ML

Only null left
Add to Cart
Add to Cart

Specifications

CAS 7132-64-1
Density 0.865 g/mL at 20°C; 0.862 g/mL (at 25°C)
Shipping Condition Ambient
Physical Form Neat
Linear Formula CH3(CH2)13 CO2 CH3
MDL Number MFCD00008989
Melting Point 18°C to 19°C
Boiling Point 141°C to 142°C (3 mmHg)
Synonym Pentadecanoic acid methyl ester
Type Analytical Standard
Quantity 25 mL
Formula Weight 256.42
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.