Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Pentafluoropropionate 98.0+%, TCI America™
SDP

Catalog No. P135225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P135225G 25 g
P13525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P135225G Supplier TCI America Supplier No. P135225G

Chemical Identifiers

CAS 378-75-6
Molecular Formula C4H3F5O2
Molecular Weight (g/mol) 178.058
MDL Number MFCD00000430
InChI Key JMKJCPUVEMZGEC-UHFFFAOYSA-N
Synonym methyl pentafluoropropionate, methyl perfluoropropionate, methylpentafluoropropionate, pentafluoropropionic acid methyl ester, propanoic acid, pentafluoro-, methyl ester, methyl pentafluoropropanoate, perfluoropropionic acid, methyl ester, pentafluoropropanoic acid methyl ester, 2,2,3,3,3-pentafluoro-propionic acid methyl ester, methylester kyseliny pentafluorpropionove
PubChem CID 9783
IUPAC Name methyl 2,2,3,3,3-pentafluoropropanoate
SMILES COC(=O)C(C(F)(F)F)(F)F

Specifications

Color Colorless
Boiling Point 60°C
Quantity 25 g
UN Number 3272
Formula Weight 178.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Pentafluoropropionate
RTECSNumber UF6475500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.