Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl propionate, 99%

Catalog No. p-7022257
Click to view available options
Quantity:
100 mL
500 mL
2500 mL

Catalog No. AAA14944AE

Supplier: Thermo Scientific Chemicals A14944AE

Only null left
Add to Cart

CAS: 554-12-1 | C4H8O2 | 88.106 g/mol

It is commonly used in organic synthesis. It undergoes vapor-phase aldol condensation with formaldehyde to form methyl methacrylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is commonly used in organic synthesis. It undergoes vapor-phase aldol condensation with formaldehyde to form methyl methacrylate.

Solubility
Soluble in water (5g/L).

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 554-12-1
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00009306
InChI Key RJUFJBKOKNCXHH-UHFFFAOYSA-N
Synonym methyl propionate, propanoic acid, methyl ester, methyl propylate, methylpropionate, propionic acid, methyl ester, propionate de methyle, propanoic acid methyl ester, fema number 2742, propionic acid methyl ester, methyl propionate natural
PubChem CID 11124
IUPAC Name methyl propanoate
SMILES CCC(=O)OC

Specifications

Melting Point °C to 88°C
Density 0.915
Boiling Point 78°C to 80°C
Flash Point -2°C (28°F)
Assay Percent Range 99%
Refractive Index 1.377
Quantity 100 mL
UN Number UN1248
Beilstein 1737628
Merck Index 14,6112
Solubility Information Soluble in water (5g/L).
Formula Weight 88.11
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl Propionate
Show More Show Less
Hazard Category H225-H315-H319-H332
Hazard Statement GHS H Statement
H225-H332
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: METHYL PROPIONATE
EINECSNumber 209-060-4
RTECSNumber UF5970000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.