Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Propionate, 99+%, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
Packaging:
Glass bottle
554-12-1
C4H8O2
88.11
MFCD00009306
RJUFJBKOKNCXHH-UHFFFAOYSA-N
methyl propionate,propanoic acid, methyl ester,methyl propylate,methylpropionate,propionic acid, methyl ester,propionate de methyle,propanoic acid methyl ester,fema number 2742,propionic acid methyl ester,methyl propionate natural
11124
methyl propanoate
CCC(=O)OC
554-12-1
C4H8O2
88.11
MFCD00009306
RJUFJBKOKNCXHH-UHFFFAOYSA-N
methyl propionate,propanoic acid, methyl ester,methyl propylate,methylpropionate,propionic acid, methyl ester,propionate de methyle,propanoic acid methyl ester,fema number 2742,propionic acid methyl ester,methyl propionate natural
11124
methyl propanoate
CCC(=O)OC
Melting Point -88°C
Boiling Point 78°C to 79°C
Flash Point -2°C
Specific Gravity 0.915
Refractive Index 1.377
Density 0.9150g/mL
Infrared Spectrum Authentic
Water 0.3% max. (K.F.)
Color Colorless
Quantity 1L
Acidity 1mg KOH/g max.
Packaging Glass bottle
Assay Percent Range 99+%
Linear Formula C2H5CO2CH3
Beilstein 02, 239
Merck Index 15, 6185
Solubility Information Solubility in water: 5g/L water (20°C). Other solubilities: miscible with alcohol and ether,soluble in most common organic solvents
Formula Weight 88.11
Physical Form Liquid
Percent Purity ≥99.0%
Chemical Name or Material Methyl propionate, 99+%
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Highly flammable liquid and vapor.

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 209-060-4

RTECSNumber : UF5970000

TSCA : TSCA