Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methyl Purple Indicator, in dilute IPA (15% v/v), Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber Glass Bottle
Specifications
Specifications
Chemical Name or Material | Methyl Purple Indicator |
CAS | 7732-18-5, 67-63-0, 845-10-3, 3844-45-9 |
CAS Min % | 88.0, 10.26, 0.06, 0.03 |
CAS Max % | 91.0, 10.68, 0.07, 0.03 |
Concentration | 15% (v/v) |
Grade | Indicator |
Packaging | Amber Glass Bottle |
Solvent or Matrix | Dilute IPA |
Color | Dark Green |
pH | >5.4 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction