Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methyl Red, ACS Reagent (Free Acid) (Acid Red 2 Not water soluble), Reagents
Click to view available options
Quantity:
10 g
25 g
Specifications
Specifications
Grade | ACS |
Packaging | Glass Bottle |
Specific Gravity | 0.79 |
Color | Brown to Red |
Quantity | 25 g |
Physical Form | Solid |
Chemical Name or Material | Methyl Red, ACS Reagent (Free Acid) (Acid Red 2 Not water soluble) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction