Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl red, For ACS analysis, MP Biomedicals™

Catalog No. ICN15254580 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15254580 100 g
ICN15254525 25 g
ICN15254583 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15254580 Supplier MP Biomedicals, Inc Supplier No. 0215254580
Add to Cart
Add to Cart

Chemical Identifiers

CAS 493-52-7
Molecular Formula C15H15N3O2
Molecular Weight (g/mol) 269.304
InChI Key CEQFOVLGLXCDCX-UHFFFAOYSA-N
Synonym 2-(4-Dimethylaminophenylazo)benzoic acid, 4-Dimethylaminoazobenzene-2'-carboxylic acid
PubChem CID 10303
IUPAC Name 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O

Specifications

Formula Weight 269.3
Color Red
Quantity 100 g
Physical Form Powder
Chemical Name or Material Methyl red, For ACS analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.