Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methyl Red Indicator, 0.02% (w/v), Ricca Chemical

Click to view available options
Quantity:
120 mL
500 mL
1 L
Packaging:
Natural Poly Bottle
Specifications
Specifications
Chemical Name or Material | Methyl Red Indicator |
CAS | 845-10-3, 7732-18-5 |
Concentration | 0.02% (w/v) |
Percent Purity | 0.02% W/V |
Grade | Indicator |
Packaging | Natural Poly Bottle |
Solvent or Matrix | Aqueous Solution |
Color | Orange to Red |
pH | 5 |
Quantity | 1 L |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction