Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methyl Red Indicator, 0.02% (w/v) in Methanol, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle
Specifications
Specifications
Chemical Name or Material | Methyl Red Indicator |
CAS | 67-56-1, 493-52-7 |
Percent Purity | 0.02% W/V |
Grade | Indicator |
Concentration or Composition (by Analyte or Components) | 0.02% (w/v) in Methanol |
Packaging | Natural Poly Bottle |
Color | Red |
Quantity | 500 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction