Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Methyl Red Indicator
SCI_ED

Catalog No. s25434 Shop All Fisherbrand Products
Click to view available options
Quantity:
10 g
25 g
Packaging:
Solid
This item is not returnable. View return policy
This item is not returnable. View return policy
pH 4.8-6.0 (red-yellow)

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 845-10-3
Molecular Formula C15H14N3NaO2
Molecular Weight (g/mol) 291.286
InChI Key GNTPCYMJCJNRQB-UHFFFAOYSA-M
Synonym Sodium Salt, Methyl Red In Alcohol
PubChem CID 4465632
IUPAC Name sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+]

Specifications

Linear Formula C15H14N3O2Na
Formula Weight 291.28
Packaging Solid
Density 0.78g/mL
Color Orange
Quantity 10 g
Chemical Name or Material Methyl Red Indicator
ShelfLife 5 years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.