Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Red Indicator, Reag. Ph. Eur., Solstice

Catalog No. 60046143 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-143 100 g Glass Bottle
60-046-144 25 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-046-143 Supplier Solstice Supplier No. 32654100G
Add to Cart
Add to Cart

Reag. Ph. Eur., acid-base indicator

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 493-52-7
Molecular Formula C15H15N3O2
Molecular Weight (g/mol) 269.304
MDL Number MFCD00002425
InChI Key CEQFOVLGLXCDCX-UHFFFAOYSA-N
Synonym Acid Red 2
PubChem CID 10303
IUPAC Name 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O

Specifications

Melting Point 179°C to 182°C
Linear Formula (CH3)2NC6H4N=NC6H4CO2H
UN Number NONH for all modes of transport
Beilstein 750102
Solubility Information Slightly soluble in water
Formula Weight 269.3g/mol
Packaging Glass Bottle
Suitability Complies for sensitivity
Visual Transition Interval 4.4 to 6.0
Density 0.989g/cm3
Color Violet
Quantity 100 g
Physical Form Powder
Chemical Name or Material Methyl Red, Reag. Ph. Eur.
Show More Show Less
Hazard Statement P273-P391-P501
Precautionary Statement H411
EINECSNumber 207-776-1
RTECSNumber DG8960000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.