Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methyl Red Mixed Solution R, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Amber Glass Bottle
Specifications
Specifications
| Chemical Name or Material | Methyl Red Mixed Solution |
| Name Note | Reagent |
| CAS | 64-17-5, 67-56-1, 493-52-7, 7220-79-3 |
| CAS Min % | 98.0, 0.12, 0.06 |
| CAS Max % | 100.0, 0.13, 0.06 |
| Grade | Indicator |
| Packaging | Amber Glass Bottle |
| Color | Dark Green |
| Quantity | 120 mL |
| Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction