Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methyl Red, Sodium Salt (Certified ACS), Fisher Chemical
Shop All Fisherbrand Products
Click to view available options
Quantity:
10 g
Specifications
Specifications
Melting Point | 180°C |
Clarity | Pass Test |
Linear Formula | 4-(CH3)2NC6H4-N:N-2-(CO2Na)C6H4 |
Identification | Pass Test |
Formula Weight | 291.28 |
Grade | Certified ACS |
Packaging | Glass Bottle |
Visual Transition Interval | Pass Test |
Color | Orange |
Quantity | 10 g |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction