Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl (S)-2-Isocyanato-3-phenylpropionate 97.0+%, TCI America™
SDP

Catalog No. I04725G
Click to view available options
Quantity:
5 g

Chemical Identifiers

CAS 40203-94-9
Molecular Formula C11H11NO3
Molecular Weight (g/mol) 205.21
MDL Number MFCD00191535
InChI Key JOTMMIYKEOOTNZ-UHFFFAOYNA-N
Synonym (S)-2-Isocyanato-3-phenylpropionic Acid Methyl Ester
PubChem CID 1514293
IUPAC Name methyl 2-isocyanato-3-phenylpropanoate
SMILES COC(=O)C(CC1=CC=CC=C1)N=C=O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 205.21
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Methyl (S)-2-Isocyanato-3-phenylpropionate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.