Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl (S)-(-)-2-Isocyanato-4-methylvalerate 98.0+%, TCI America™
SDP

Catalog No. I046725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I046725G 25 g
I04675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I046725G Supplier TCI America Supplier No. I046725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 39570-63-3
Molecular Formula C8H13NO3
Molecular Weight (g/mol) 171.196
MDL Number MFCD00191532
InChI Key UWFLIWPVRTVDNO-ZETCQYMHSA-N
Synonym (S)-(-)-2-Isocyanato-4-methylvaleric Acid Methyl Ester
PubChem CID 7019163
IUPAC Name methyl (2S)-2-isocyanato-4-methylpentanoate
SMILES CC(C)CC(C(=O)OC)N=C=O

Specifications

Color Yellow
Boiling Point 74°C
Quantity 25 g
UN Number 2206
Formula Weight 171.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl (S)-(-)-2-Isocyanato-4-methylvalerate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.