Learn More
Methyl (S)-(+)-3-hydroxy-2-methylpropionate, 98%
Description
Methyl (S)-(+)-3-hydroxy-2-methylpropionate is used as a starting material for the synthesis of dictyostatin, discodermolide and spongidepsin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Colorless |
| Density | 1.07 g/mL |
| Boiling Point | 74°C (10 mmHg) |
| Flash Point | 81°C (178°F) |
| Refractive Index | 1.425 |
| Quantity | 10 g |
| Beilstein | 1720830 |
| Solubility Information | Insoluble in water. Soluble in methanol. |
| Optical Rotation | +27° (c=4 in methanol) |
| Formula Weight | 118.13 |
| Show More |
Safety and Handling
| Hazard Category | H227-H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335-H227 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Combustible liquid. |
| Precautionary Statement | P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.