Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl (S)-(+)-3-hydroxy-2-methylpropionate, 98%

Catalog No. AA4448304
Click to view available options
Quantity:
2 g
10 g

CAS: 80657-57-4 | C5H10O3 | 118.13 g/mol

Methyl (S)-(+)-3-hydroxy-2-methylpropionate is used as a starting material for the synthesis of dictyostatin, discodermolide and spongidepsin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80657-57-4
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.13
MDL Number MFCD00064520
InChI Key ATCCIZURPPEVIZ-UHFFFAOYNA-N
Synonym s-methyl 3-hydroxy-2-methylpropanoate, methyl s-+-3-hydroxy-2-methylpropionate, s-+-3-hydroxy-2-methylpropionic acid methyl ester, methyl 2s-3-hydroxy-2-methylpropanoate, 2-methyl-3-hydroxypropanoic acid, propanoic acid, 3-hydroxy-2-methyl-, methyl ester, 2s, s-+-methyl 3-hydroxy-2-methylpropionate, s-3-hydroxy-2-methylpropionic acid methyl ester, +-methyl l-beta-hydroxyisobutyrate, methyl 3-hydroxy-2-methylpropanoate #
PubChem CID 10192563
IUPAC Name methyl (2S)-3-hydroxy-2-methylpropanoate
SMILES COC(=O)C(C)CO

Specifications

Color Colorless
Density 1.07 g/mL
Boiling Point 74°C (10 mmHg)
Flash Point 81°C (178°F)
Refractive Index 1.425
Quantity 2 g
Beilstein 1720830
Solubility Information Insoluble in water. Soluble in methanol.
Optical Rotation +27° (c=4 in methanol)
Formula Weight 118.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl (S)- (+)-3-hydroxy-2-methylpropionate
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.