Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methyl (S)-(-)-lactate, 97%
Click to view available options
Quantity:
25 g
100 g
Description
It is used in pharmaceutical industry, agricultural fields and is also used as a solvent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in pharmaceutical industry, agricultural fields and is also used as a solvent.
Solubility
Miscible. Hydrolyzes in water.
Notes
Store at room temperature. Incompatible with oxidizing agents.
It is used in pharmaceutical industry, agricultural fields and is also used as a solvent.
Solubility
Miscible. Hydrolyzes in water.
Notes
Store at room temperature. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | -44°C |
| Density | 1.092 |
| Boiling Point | 135°C to 138°C |
| Flash Point | 49°C (120°F) |
| Refractive Index | 1.414 |
| Quantity | 25 g |
| UN Number | UN3272 |
| Beilstein | 1720587 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Miscible. Hydrolyzes in water. |
| Show More |
Safety and Handling
| Hazard Category | H226-H319-H335 |
| Hazard Statement | GHS H Statement H226-H319-H335 Flammable liquid and vapour. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S. |
| EINECSNumber | 248-704-9 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction