Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Salicylate, NF, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18604456 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Steel Can
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-456 20 L Steel Can
18-604-454 500 mL Steel Can
18-604-455 4 L Steel Can
18-604-457 200 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-604-456 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. ME17020LTMT

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

ME170, 119-36-8, C8H8O3

Spectrum™ Chemical Methyl Salicylate, NF is used to treat minor aches and pains of the muscles and joints. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 119-36-8
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
InChI Key OSWPMRLSEDHDFF-UHFFFAOYSA-N
IUPAC Name methyl 2-hydroxybenzoate
SMILES COC(=O)C1=CC=CC=C1O

Specifications

CAS Max % 100%
Packaging Steel Can
Quantity 20 L
Percent Purity 98 to 102%
Grade NF
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.