Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Salicylate (Reagent), Fisher Chemical

Catalog No. o3695500 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL
Packaging:
Glass Bottle

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-36-8
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00002214
InChI Key OSWPMRLSEDHDFF-UHFFFAOYSA-N
Synonym methyl salicylate, wintergreen oil, gaultheria oil, betula oil, teaberry oil, sweet birch oil, oil of wintergreen, analgit, spicewood oil, 2-hydroxybenzoic acid methyl ester
PubChem CID 4133
ChEBI CHEBI:31832
IUPAC Name methyl 2-hydroxybenzoate
SMILES COC(=O)C1=CC=CC=C1O

Specifications

Melting Point -8.6°C
Color Yellow to Orange
Density 1.18g/mL
Boiling Point 223.3°C
Assay Percent Range ≥99 %
Packaging Glass Bottle
Linear Formula 2-(CO2CH3)C6H4OH
Quantity 500 mL
Vapor Pressure 00.1 mmHg at 20°C
Formula Weight 152.15
Percent Purity ≥99%
Grade Reagent
Physical Form Liquid
Chemical Name or Material Methyl Salicylate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.