Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl sulfone, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
500g
2.5kg
Packaging:
Glass Bottle
Plastic bottle
67-71-0
C2H6O2S
94.13
HHVIBTZHLRERCL-UHFFFAOYSA-N
dimethyl sulfone,methyl sulfone,sulfonylbismethane,dimethylsulfone,dimethyl sulphone,methane, sulfonylbis,sulphonylbismethane,methylsulfone,methanesulfonylmethane,sulfonyldimethane
6213
CHEBI:9349
methylsulfonylmethane
CS(=O)(=O)C
67-71-0
C2H6O2S
94.13
HHVIBTZHLRERCL-UHFFFAOYSA-N
dimethyl sulfone,methyl sulfone,sulfonylbismethane,dimethylsulfone,dimethyl sulphone,methane, sulfonylbis,sulphonylbismethane,methylsulfone,methanesulfonylmethane,sulfonyldimethane
6213
CHEBI:9349
methylsulfonylmethane
CS(=O)(=O)C
Quantity 5g
Color White
Packaging Glass Bottle
Melting Point 107.0°C to 111.0°C
Boiling Point 238.0°C
Infrared Spectrum Authentic
Flash Point 143°C
Assay Percent Range 98%
Linear Formula (CH3)2SO2
Beilstein 01,289
Fieser 01,296; 02,157
Merck Index 15,3283
Solubility Information Solubility in water: 150g/L (20°C). Other solubilities: freely soluble in methanol,ethanol and acetone,sparingly soluble in ether
Formula Weight 94.13
Physical Form Crystals or Crystalline Powder
Percent Purity 98%
Chemical Name or Material Methyl sulfone
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protec

GHS Signal Word: Warning

EINECSNumber : 200-665-9