Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl sulfoxide-d6, for NMR, with 0.03% TMS, 99.9 atom% D, ACROS Organics™

Click to view available options
Quantity:
10mL
25mL
100mL
Packaging:
Glass bottle
2206-27-1
C2H6OS
84.166
MFCD00002090
IAZDPXIOMUYVGZ-WFGJKAKNSA-N
dimethyl sulfoxide-d6,dmso-d6,dimethylsulfoxide-d6,deuterated dmso,methane-d3,sulfinylbis-9ci,methane-d3, sulfinylbis,hexadeuterodimethyl sulfoxide,methane-d3-, sulfinylbis,methanesulfinylmethyl hydrogen,dimethyl-d6 sulfoxide
75151
trideuterio(trideuteriomethylsulfinyl)methane
CS(=O)C
2206-27-1
C2H6OS
84.166
MFCD00002090
IAZDPXIOMUYVGZ-WFGJKAKNSA-N
dimethyl sulfoxide-d6,dmso-d6,dimethylsulfoxide-d6,deuterated dmso,methane-d3,sulfinylbis-9ci,methane-d3, sulfinylbis,hexadeuterodimethyl sulfoxide,methane-d3-, sulfinylbis,methanesulfinylmethyl hydrogen,dimethyl-d6 sulfoxide
75151
trideuterio(trideuteriomethylsulfinyl)methane
CS(=O)C
Color Colorless
Melting Point 20.2°C
Boiling Point 190.0°C
Viscosity 2.4 mPa.s (20°C)
Flash Point 88°C
Specific Gravity 1.19
Packaging Glass bottle
Deuterium (D) 99.85% min.
Water 0.03% max. (Coulometric)
Quantity 10mL
Density 1.1900g/mL
Beilstein 01, IV, 1279
Fieser 13,124; 16,177
Formula Weight 84.18
Physical Form Liquid
Grade NMR
Chemical Name or Material Methyl sulfoxide-d6, with 0.03% TMS, 99.9 atom%D
Show More Show Less