Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl sulfoxide-d6, for NMR, 99.8 atom % D

Catalog No. p-168097
Click to view available options
Quantity:
10 mL
25 mL
50 mL
100 mL
Packaging:
Glass Bottle
Septum Bottle

CAS: 2206-27-1 | C2H6OS | 84.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2206-27-1
Molecular Formula C2H6OS
Molecular Weight (g/mol) 84.17
MDL Number MFCD00002090
InChI Key IAZDPXIOMUYVGZ-WFGJKAKNSA-N
Synonym dimethyl sulfoxide-d6, dmso-d6, dimethylsulfoxide-d6, deuterated dmso, methane-d3,sulfinylbis-9ci, methane-d3, sulfinylbis, hexadeuterodimethyl sulfoxide, methane-d3-, sulfinylbis, methanesulfinylmethyl hydrogen, dimethyl-d6 sulfoxide
PubChem CID 75151
SMILES [2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]

Specifications

Physical Form Liquid
Color Colorless
Quantity 10 mL
Packaging Glass Bottle
Viscosity 2.4 mPa.s (20°C)
Flash Point 88°C
Specific Gravity 1.19
Density 1.1900g/mL
Deuterium (D) 99.75% min.
Water 0.03% max. (Coulometric)
Melting Point 20.2°C
Boiling Point 190.0°C
Linear Formula (CD3)2SO
Beilstein 01, IV, 1279
Fieser 13,124; 16,177
IUPAC Name trideuterio(trideuteriomethylsulfinyl)methane
Formula Weight 84.18
Grade NMR
Chemical Name or Material Methyl sulfoxide-d6, 99.8 atom % D
Show More Show Less
Hazard Category HYGROSCOPIC
EINECSNumber 218-617-0
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.