Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl thiazole-2-carboxylate, 95%

Catalog No. AAH63496MD
Click to view available options
Quantity:
250 mg
1 g
5 g

CAS: 55842-56-3 | C5H5NO2S | 143.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55842-56-3
Molecular Formula C5H5NO2S
Molecular Weight (g/mol) 143.16
MDL Number MFCD18075580
InChI Key YRMZUOPMHMPNPG-UHFFFAOYSA-N
PubChem CID 7567660
IUPAC Name methyl 1,3-thiazole-2-carboxylate
SMILES COC(=O)C1=NC=CS1

Specifications

Quantity 250 mg
Synonym methyl thiazole-2-carboxylate, methyl 2-thiazolecarboxylate, 2-thiazolecarboxylic acid methyl ester, methyl2-thiazolecarboxylate
Formula Weight 143.16
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Methyl thiazole-2-carboxylate
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.