Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl thiocyanate, 99%

Catalog No. p-7030311
Click to view available options
Quantity:
5 mL
25 mL
100 mL

CAS: 556-64-9 | C2H3NS | 73.113 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 556-64-9
Molecular Formula C2H3NS
Molecular Weight (g/mol) 73.113
MDL Number MFCD00001830
InChI Key VYHVQEYOFIYNJP-UHFFFAOYSA-N
Synonym methyl sulfocyanate, methylrhodanid, thiocyanic acid, methyl ester, methyl rhodanate, methylthiokyanat, methane, thiocyanato, thiocyanatomethane, methylthiocyanate, thiocyanic acid methyl ester, ch3scn
PubChem CID 11168
ChEBI CHEBI:61112
IUPAC Name methyl thiocyanate
SMILES CSC#N

Specifications

Melting Point -5°C
Density 1.068
Boiling Point 131°C
Flash Point 38°C (100°F)
Odor Pungent
Refractive Index 1.468
Quantity 5 mL
UN Number UN2929
Sensitivity Moisture sensitive
Formula Weight 73.11
Percent Purity 99%
Chemical Name or Material Methyl thiocyanate
Show More Show Less
Hazard Category H226-H301+H311+H331-H315-H319
Hazard Statement GHS H Statement
H300-H330-H226-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
EINECSNumber 209-134-6
RTECSNumber XL1575000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.