Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl tiglate, 98%

Catalog No. p-7081793
Click to view available options
Quantity:
50 g
250 g

CAS: 6622-76-0 | C6H10O2 | 114.14 g/mol

It is employed as a perfuming agent in cosmetics industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a perfuming agent in cosmetics industry.

Solubility
Soluble in alcohol, water 3813 mg/L @ 25°C (est).

Notes
Store away from oxidizing agents. Keep the container tightly closed. Store in cool, dry conditions in well ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6622-76-0
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD00016654
InChI Key YYJWBYNQJLBIGS-PLNGDYQASA-N
Synonym methyl tiglate, tiglic acid methyl ester, methyl 2-methylbut-2-enoate, methyl 2-methyl-2-butenoate, methyl e-2-methylcrotonate, methyl 2-methylcrotonate, methyl alpha-methylcrotonate, methyl trans-2-methylcrotonate, 2-butenoic acid, 2-methyl-, methyl ester, e, crotonic acid, 2-methyl-, methyl ester, e
PubChem CID 5323652
IUPAC Name methyl (E)-2-methylbut-2-enoate
SMILES COC(=O)C(\C)=C/C

Specifications

Density 0.95
Boiling Point 138°C to 139°C
Flash Point 39°C (102°F)
Refractive Index 1.437
Quantity 50 g
UN Number UN3272
Beilstein 1720455
Merck Index 14,9433
Solubility Information Soluble in alcohol,water 3813mg/L @ 25°C (est).
Formula Weight 114.15
Percent Purity 98%
Chemical Name or Material Methyl tiglate
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 229-575-8
RTECSNumber EM9254600
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.