Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Tridecafluoroheptanoate 96.0+%, TCI America™
SDP

Catalog No. M191425G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 14312-89-1
Molecular Formula C8H3F13O2
Molecular Weight (g/mol) 378.089
MDL Number MFCD00153212
InChI Key JHROQORAJUWVCD-UHFFFAOYSA-N
Synonym Methyl Perfluoroheptanoate, Tridecafluoroheptanoic Acid Methyl Ester, Perfluoroheptanoic Acid Methyl Ester
PubChem CID 526471
IUPAC Name methyl 2,2,3,3,4,4,5,5,6,6,7,7,7-tridecafluoroheptanoate
SMILES COC(=O)C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Color Colorless
Boiling Point 141°C
Quantity 25 g
UN Number 1993
Formula Weight 378.09
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Tridecafluoroheptanoate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.