Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl Trifluoromethanesulfonate

Catalog No. 16428350G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Click to view available options
Quantity:
10 g
50 g
250 g
Danger

Catalog No. 16-428-350G

Supplier: Sigma Aldrich 16428350G

Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Methyl trifluoromethanesulfonate is a strong methylating reagent, commonly used for the pre-methylation of polysaccharides under mild basic conditions.Methyl trifluoromethanesulfonate can be used as a methylation reagent:In the determination of polysulfides, zerovalent sulfur in sulfide-rich water wells, and polysulfide species in electrolyte of a lithium-sulfur battery using chromatography-based techniques.In reactions with potassium enolates.For the conversion of amines to methyl ammonium triflates.

Specifications

CAS 333-27-7
Density 1.45 g/mL at 25°C
Boiling Point 94°C to 99°C
Molecular Formula C2H3F3O3S
Refractive Index n20/D 1.326
Linear Formula CF3SO2OCH3
MDL Number MFCD00000409
Quantity 50 g
Synonym Methyl triflate
Molecular Weight (g/mol) 164.1 g/mol
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.