Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl (triphenylphosphoranylidene)acetate, 98%

Catalog No. AC169561000
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle

CAS: 2605-67-6 | C21H19O2P | 334.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2605-67-6
Molecular Formula C21H19O2P
Molecular Weight (g/mol) 334.36
MDL Number MFCD00008455
InChI Key NTNUDYROPUKXNA-UHFFFAOYSA-N
Synonym methyl triphenylphosphoranylidene acetate, carbomethoxymethylene triphenylphosphorane, methyl 2-triphenylphosphoranylidene acetate, acetic acid, triphenylphosphoranylidene-, methyl ester, methyl triphenylphosphoranylideneacetate, triphenylphosphoranylidene acetic acid methyl ester, triphenyl carbomethoxymethylene phosphorane, methoxycarbonylmethylene triphenylphosphorane, acetic acid, 2-triphenylphosphoranylidene-, methyl ester
PubChem CID 17453
IUPAC Name methyl 2-(triphenyl-$l^{5}-phosphanylidene)acetate
SMILES COC(=O)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 164.0°C to 169.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 2% max. triphenylphosphine+ triphenylphosphine oxide (HPLC)
Packaging Glass bottle
Linear Formula (C6H5)3P=CHCO2CH3
Quantity 100 g
Fieser 01,112; 01,1081; 08,456; 09,426; 10,363; 13,277
Solubility Information Solubility in water: insoluble
Formula Weight 334.34
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Methyl (triphenylphosphoranylidene)acetate
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 220-018-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.