Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molybdic acid, ACS reagent

Catalog No. AC423980025
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle

CAS: 7782-91-4 | H2MoO4 | 161.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-91-4
Molecular Formula H2MoO4
Molecular Weight (g/mol) 161.96
MDL Number MFCD00036287
InChI Key VLAPMBHFAWRUQP-UHFFFAOYSA-L
Synonym molybdic acid, molybdic vi acid, unii-i96n991j1n, dihydroxy dioxo molybdenum, molybdic acid acs, dihydroxy-dioxomolybdenum, molybdate moo42-, dihydrogen, t-4, dihydroxidodioxidomolybdenum, aronis24112, moo2 oh 2
PubChem CID 82208
ChEBI CHEBI:25371
IUPAC Name dihydroxy(dioxo)molybdenum
SMILES O[Mo](O)(=O)=O

Specifications

Color White
Physical Form Crystalline Powder and/or Crystals
Quantity 2.5 kg
Merck Index 15,6326
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in nh4oh, h2so4 and fixed alkalies
Formula Weight 161.96
Percent Purity ≥85.0% (MoO3)
Grade ACS Reagent
Packaging Plastic bottle
Specific Gravity 3.1
Density 3.1000g/mL
Chemical Name or Material Molybdic acid, For Analysis ACS
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause damage to organs through prolonged or repeated exposure.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear eye protection/face protection.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/
EINECSNumber 231-970-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.