Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl trisulfide, 98+%

Catalog No. ac415030050
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 3658-80-8 | C2H6S3 | 126.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3658-80-8
Molecular Formula C2H6S3
Molecular Weight (g/mol) 126.27
MDL Number MFCD00039808
InChI Key YWHLKYXPLRWGSE-UHFFFAOYSA-N
Synonym dimethyl trisulfide, trisulfide, dimethyl, methyl trisulfide, 2,3,4-trithiapentane, dimethyl trisulphide, dimethyltrisulfide, methyltrisulfanyl methane, dimethyltrisulfane, unii-3e691t3nl1, dmts
PubChem CID 19310
ChEBI CHEBI:4614
IUPAC Name (methyltrisulfanyl)methane
SMILES CSSSC

Specifications

Melting Point -68.0°C
Color Yellow
Density 1.1900g/mL
Boiling Point 165.0°C to 175.0°C
Flash Point 52°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.5995 to 1.6025
Linear Formula CH3SSSCH3
Quantity 5 g
Beilstein 01, IV, 1283
Specific Gravity 1.19
Formula Weight 126.27
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Dimethyl trisulfide, 98+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Call a POISON CENTER or doctor/physician if you feel unwell.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/prot
EINECSNumber 222-910-9
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.