Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl undecanoate, 99%

Catalog No. p-7044340
Click to view available options
Quantity:
5 g
25 g

CAS: 1731-86-8 | C12H24O2 | 200.322 g/mol

Methyl undecanoate is used as plasticizer in polymers. It is used as fragrance agent in aroma and flavor Chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl undecanoate is used as plasticizer in polymers. It is used as fragrance agent in aroma and flavor Chemicals.

Solubility
Soluble in ethanol, and ether. Insoluble in water.

Notes
Store at -20°C. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1731-86-8
Molecular Formula C12H24O2
Molecular Weight (g/mol) 200.322
MDL Number MFCD00008957
InChI Key XPQPWPZFBULGKT-UHFFFAOYSA-N
Synonym undecanoic acid methyl ester, undecanoic acid, methyl ester, methyl n-undecanoate, unii-k67yz97w6c, undecanoic acid methylester, n-undecanoic acid methyl ester, methyl undecanoate, acmc-209e5z, undecanoic acid, methylester, 3-02-00-00858 beilstein handbook reference
PubChem CID 15607
ChEBI CHEBI:87527
IUPAC Name methyl undecanoate
SMILES CCCCCCCCCCC(=O)OC

Specifications

Melting Point -10°C
Density 0.871
Boiling Point 246°C to 248°C
Flash Point 109°C (228°F)
Refractive Index 1.429
Quantity 5 g
Beilstein 1762129
Solubility Information Soluble in ethanol,and ether. Insoluble in water.
Percent Purity 99%
Chemical Name or Material Methyl undecanoate
Hazard Category H315-H318-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
EINECSNumber 217-053-2
RTECSNumber YQ2573000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.