Alfa Aesar™ Methyl valerate, 99%

Manufacturer: Alfa Aesar™ B2482836

Quantity 500g

 View more versions of this product

Catalog No. AAB2482836

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 624-24-8
    Molecular Formula CH3(CH2)3CO2CH3
    Formula Weight 116.16
    MDL Number MFCD00009478
    UN Number UN3272
    Beilstein 1741905
    Synonym Methyl pentanoate; Valeric acid methyl ester
    Chemical Name or Material Methyl valerate

    Specifications

    Formula Weight 116.16
    TSCA TSCA
    Molecular Formula CH3(CH2)3CO2CH3
    MDL Number MFCD00009478
    UN Number UN3272
    Synonym Methyl pentanoate; Valeric acid methyl ester
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H332
    Highly flammable liquid and vapour.
    Harmful if inhaled.
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number YV7750500
    Flash Point 22°C (71°F)
    Boiling Point 126° to 128°C
    Melting Point −91°C
    Quantity 500g
    CAS 624-24-8
    Beilstein 1741905
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ESTERS, N.O.S.
    Chemical Name or Material Methyl valerate
    Assay 99%

    GHS H Statement
    H225-H332
    Highly flammable liquid and vapour.
    Harmful if inhaled.

    GHS P Statement
    P210-P261-P280-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit